Learn More
Advanced Instruments Inc MVS BUNDLE DM
SDP
Catalog No. NC2259404
Encompass
Advanced/800TSNB Reader-For verification of 1, 2, 4, 6,8, 12, and 96 tips with 96-wellplates and 1, 8, 12, 16, 24,96 and 384 tips with 384-wellplates Equipment Packageincludes: •Certified Artel800TNSB Plate Reader•Calibrator Plate/Carry Case•Titer Plate Shaker•Bar Code Reader •PortableComputer•Data Manager ADVANCED•Mobile Work Station •DigitalUser GuideConsumables•Customized MVSConsumables for start up,
Catalog No. NC2259404 Supplier Advanced Instruments Inc Supplier No. MVS540
Add to Cart
Add to Cart
Request a Quote
Advanced/800TSNB Reader-For verification of 1, 2, 4, 6,8, 12, and 96 tips with 96-wellplates and 1, 8, 12, 16, 24,96 and 384 tips with 384-wellplates Equipment Packageincludes: •Certified Artel800TNSB Plate Reader•Calibrator Plate/Carry Case•Titer Plate Shaker•Bar Code Reader •PortableComputer•Data Manager ADVANCED•Mobile Work Station •DigitalUser GuideConsumables•Customized MVSConsumables for start up,