Learn More
Advanced Instruments Inc MVS BUNDLE DM STANDARD/800TSNB
SDP
Catalog No. NC2035533
Encompass
MVS Bundle DM Standard/800TSNB Reader - For verification of 1, 2, 4, 6, 8, 12, and 96 tips with 96-well plates Equipment
Catalog No. NC2035533 Supplier Advanced Instruments Inc Supplier No. MVS500
Add to Cart
Add to Cart
Request a Quote
MVS Bundle DM Standard/800TSNB Reader - For verification of 1, 2, 4, 6, 8, 12, and 96 tips with 96-well plates Equipment