Learn More
Advanced Instruments Inc MVS DMSO QualAssure D Solution, Accurate Verification of Liquid Transfers, Traceable to International Standards, 120 mL
SDP
Supplier:  Advanced Instruments Inc MVS218
Encompass
The Advanced Instruments MVS DMSO QualAssure D Solution is a specialized liquid handling verification solution, optimizing DMSO-based transfers for accurate and precise delivery in multichannel systems. Traceable to international standards, it simplifies calibration and ensures nL precision, benefiting high-throughput applications like compound management and drug discovery. Its ease of use and rapid verification (under 10 min) differentiates it, providing an essential tool for laboratories focused on accuracy and efficiency.
Catalog No. NC0637112