Learn More
Advanced Instruments Inc MVS-207 Aqueous QualAssure Solution, Automated Liquid Handler Verification, Compliant with ISO Standards, 120 mL
SDP
Catalog No. NC0901525
Encompass
The Advanced Instruments MVS-207 Aqueous QualAssure solution is an essential consumable for verifying the accuracy of automated liquid handlers and multichannel pipettes. This solution enhances performance by mimicking aqueous-based assay reagents, ensuring precise liquid handling with reduced variability. It adheres to international regulatory standards, making it ideal for laboratories aiming to maintain data integrity and optimize results quickly.
Catalog No. NC0901525 Supplier Advanced Instruments Inc Supplier No. MVS207
The Advanced Instruments MVS-207 Aqueous QualAssure solution is an essential consumable for verifying the accuracy of automated liquid handlers and multichannel pipettes. This solution enhances performance by mimicking aqueous-based assay reagents, ensuring precise liquid handling with reduced variability. It adheres to international regulatory standards, making it ideal for laboratories aiming to maintain data integrity and optimize results quickly.