Learn More
Advanced Instruments Inc MVS VERIFICATION PLATES-N 25BX
SDP
Catalog No. NC9269159
Encompass
MVS Verification Plate; 96-well; Optical quality; Flat bottom plates which are dimensionally characterized and bar coded with lot number and a unique identifier; 25/PK
Catalog No. NC9269159 Supplier Advanced Instruments Inc Supplier No. MVS230
MVS Verification Plate; 96-well; Optical quality; Flat bottom plates which are dimensionally characterized and bar coded with lot number and a unique identifier; 25/PK