Learn More
Medchemexpress LLC HY-120111 10mg Medchemexpress, MW-150 CAS:1628502-91-9 Purity:>98%
SDP
Catalog No. 501969578
Encompass_Preferred
Medchemexpress, HY-120111 10mg MW-150 CAS:1628502-91-9 MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9578 Supplier Medchemexpress LLC Supplier No. HY12011110MG
Medchemexpress, HY-120111 10mg MW-150 CAS:1628502-91-9 MW150 (MW01-18-150SRM) is a selective, CNS penetrant, and orally active inhibitor of p38α MAPK with a Ki of 101 nM. MW-150 inhibits the ability of the endogenous p38α MAPK to phosphorylate an endogenous substrate MK2 in activated glia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.