Learn More
Cell Signaling Technology Biotinylated Protein Ladder Detection Pack, 9 kDa to 200 kDa, 3,250 μL
SDP
Catalog No. 50193777
Encompass
The Biotinylated Protein Ladder Detection Pack is designed for molecular weight ladder detection on Western blots using horseradish peroxidase (HRP)-based detection systems. The pack contains a Biotinylated Protein Ladder, consisting of purified proteins covalently coupled to biotin that resolve into 10 bands with a size range of 9–200 kDa, and an Anti-biotin, HRP-linked Antibody for detection.
  • Optimized for chemiluminescent Western detection procedures.
  • Molecular weight range of 9–200 kDa.
  • Comprises 10 distinct protein bands.
  • Includes Biotinylated Protein Ladder and Anti-biotin, HRP-linked Antibody.
  • Validated for Western blotting applications.
Catalog No. 50-193-777 Supplier Cell Signaling Technology Supplier No. 7727L
The Biotinylated Protein Ladder Detection Pack is designed for molecular weight ladder detection on Western blots using horseradish peroxidase (HRP)-based detection systems. The pack contains a Biotinylated Protein Ladder, consisting of purified proteins covalently coupled to biotin that resolve into 10 bands with a size range of 9–200 kDa, and an Anti-biotin, HRP-linked Antibody for detection.
  • Optimized for chemiluminescent Western detection procedures.
  • Molecular weight range of 9–200 kDa.
  • Comprises 10 distinct protein bands.
  • Includes Biotinylated Protein Ladder and Anti-biotin, HRP-linked Antibody.
  • Validated for Western blotting applications.