Learn More
New England Biolabs, Inc. MwoI – 2500 units
SDP
Catalog No. 50812269
Encompass
A restriction endonuclease that recognizes the sequence GCNN_NNN^NNGC.
Catalog No. 50-812-269 Supplier New England Biolabs, Inc. Supplier No. R0573L
A restriction endonuclease that recognizes the sequence GCNN_NNN^NNGC.