Learn More
Andwin Scientific MWS 14X14 CELLOPHANE SHT 50/PK
SDP
Catalog No. NC9389595
Encompass_Preferred
The cellophane sheet by Diversified Biotech is measured to have the dimensions of 14 x 14 cm. Gels in the middle of two sheets of cellophane can be dried easily without the utilization of any heat or vacuum gel dryers. This product is sold in a pack of 50.
Catalog No. NC9389595 Supplier Andwin Scientific Supplier No. AB110762
The cellophane sheet by Diversified Biotech is measured to have the dimensions of 14 x 14 cm. Gels in the middle of two sheets of cellophane can be dried easily without the utilization of any heat or vacuum gel dryers. This product is sold in a pack of 50.