Learn More
Andwin Scientific MWS CRYO BABIES 1.28X.5IN-1000
SDP
Supplier:  Andwin Scientific CRY1000PK
Encompass_Preferred
The Cryo-Babies label are resistant against most detergents and oil. The labels also combat organic solvents and caustic agents without peeling. This product measures 1.2" x 0.5". This is sold in a pack of 1000 labels.
Catalog No. NC9367574