Learn More
Andwin Scientific MWS STOUGH SPOT WH 3/8 1000/PK
SDP
Catalog No. NC9342258
Encompass_Preferred
The Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".
Catalog No. NC9342258 Supplier Andwin Scientific Supplier No. TSPOTS
The Tough-Spots labels on a sheet are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 3/8".