Learn More
Mitegen MX HP Cool Sample Base, SPINE Standard Design, For Cryogenic Sample Mounting, Box of 20
SDP
Catalog No. NC3238464
Encompass_Preferred
This MX HP Cool Sample Base, featuring a SPINE Standard Design, is an essential accessory for maintaining precise cryogenic temperatures during delicate sample mounting and manipulation procedures in crystallography workflows. Each sample base is meticulously engineered to securely hold and actively cool samples, ensuring their thermal stability and integrity before critical data collection or transfer to a cryogenic stream. The SPINE Standard Design guarantees broad compatibility with widely used sample mounting systems and beamline equipment, significantly streamlining your experimental setup. Supplied in a convenient box of 20, these cool sample bases provide ample supply for routine laboratory operations. Their precise design helps minimize thermal shock to sensitive crystals or biological specimens, directly contributing to higher quality diffraction data and reliable experimental outcomes.
Catalog No. NC3238464 Supplier Mitegen Supplier No. MXHPCSB20
This MX HP Cool Sample Base, featuring a SPINE Standard Design, is an essential accessory for maintaining precise cryogenic temperatures during delicate sample mounting and manipulation procedures in crystallography workflows. Each sample base is meticulously engineered to securely hold and actively cool samples, ensuring their thermal stability and integrity before critical data collection or transfer to a cryogenic stream. The SPINE Standard Design guarantees broad compatibility with widely used sample mounting systems and beamline equipment, significantly streamlining your experimental setup. Supplied in a convenient box of 20, these cool sample bases provide ample supply for routine laboratory operations. Their precise design helps minimize thermal shock to sensitive crystals or biological specimens, directly contributing to higher quality diffraction data and reliable experimental outcomes.