Learn More
Scilogex LLC SCI-M Microplate Mixer
Catalog No. NC1534199
Encompass_Preferred
Voltage/Frequency: 100-220V, 50/60Hz, US Plug. Speed Range: 0-1500 RPM (Single microplate), 0-1000 RPM (Double microplate). Compact universal small shaker suitable for 96 microtiter plates. Continuous operation.Stepless speed regulation of motor. External rotor brushless motor reduces operating noises and increases motor life.Long life quiet brushless motor. Accessories available for tube shaking. 2 Year Warranty
Catalog No. NC1534199 Supplier Scilogex LLC Supplier No. 822000049999
Voltage/Frequency: 100-220V, 50/60Hz, US Plug. Speed Range: 0-1500 RPM (Single microplate), 0-1000 RPM (Double microplate). Compact universal small shaker suitable for 96 microtiter plates. Continuous operation.Stepless speed regulation of motor. External rotor brushless motor reduces operating noises and increases motor life.Long life quiet brushless motor. Accessories available for tube shaking. 2 Year Warranty