Learn More
Medchemexpress LLC HY-100892 5mg Medchemexpress, MX69 CAS:1005264-47-0 Purity:>98%
SDP
Catalog No. 501931990
Encompass_Preferred
Medchemexpress, HY-100892 5mg MX69 CAS:1005264-47-0 MX69 is an inhibitor of MDM2/XIAP , used for cancer treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1990 Supplier Medchemexpress LLC Supplier No. HY1008925MG
Medchemexpress, HY-100892 5mg MX69 CAS:1005264-47-0 MX69 is an inhibitor of MDM2/XIAP , used for cancer treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.