Learn More
Kitted Lab Technologies MXC Blue 1000ml 2 Port Cap Assembly
Catalog No. NC2172958
Encompass
KLT's Patented MXC³ Blue 1000ml Media Transfer Bottle 38430 Cap corresponds with proper 1L volume bottle, allow for quick confirmation that each bioreactor experiment is set-up properly. Assembled with weldable tubing, V-cut on tube end to eliminate suction to bottle. Packaged in single-use, sterile Tyvek Pouch and positioned specifically to allow for controlled opening. Available in packages of 10, 20, or 40 caps. (US Patent Number: US 11,026,498 B2)
Catalog No. NC2172958 Supplier Kitted Lab Technologies Supplier No. KMXC100010PK
KLT's Patented MXC³ Blue 1000ml Media Transfer Bottle 38430 Cap corresponds with proper 1L volume bottle, allow for quick confirmation that each bioreactor experiment is set-up properly. Assembled with weldable tubing, V-cut on tube end to eliminate suction to bottle. Packaged in single-use, sterile Tyvek Pouch and positioned specifically to allow for controlled opening. Available in packages of 10, 20, or 40 caps. (US Patent Number: US 11,026,498 B2)