Learn More
Aviva Systems Biology Corp Mxd1 Antibody - C-terminal region (ARP37086_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37086_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mxd1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AW122478, Mad, Mad1ApplicationWB. Species reactivity: Cow, Mouse, Rabbit, Rat
Catalog No. 50-284-583