Learn More
Aviva Systems Biology Corp MXD3 Antibody - N-terminal region (ARP30089_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30089_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MXD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MXD3. Alternate names: MYX, MAD3, BHLHC13ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54120