Learn More
Aviva Systems Biology Corp MXD3 Antibody - N-terminal region (ARP39663_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39663_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MXD3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MXD3. Alternate names: MAD3, MGC2383, MYX, BHLHC13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-57128