Learn More
Aviva Systems Biology Corp MXD4 Antibody - N-terminal region (ARP32777_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32777_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MXD4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MXD4. Alternate names: MAD4, MST149, MSTP149, bHLHc12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Sheep
Catalog No. 50-014-54758