Learn More
MARSHALL SCIENTIFIC Edwards mXDS3s Dry Scroll Pump
SDP
Catalog No. NC2888792
Encompass_Preferred
The Edwards Vacuum range of small dry scroll pumps is expanding with the introduction of the compact mXDS3 and mXDS3s Dry Scroll Pumps. The new mXDS3 dry scroll pump delivers 1.77 CFM (3 m^3/h^-1) pumping speed at 50Hz or 2.06 CFM (3.5 m^3/h^-1) together with an ultimate pressure of 0.1 mbar making it well suited for backing turbomolecular pumps. The mXDS3 is the standard option supplied with no inlet valve fitted, while the mXDS3s comes factory fitted with an inlet valve featuring delay opening. The mXDS3s is also supplied with an exhaust silencer. Both offer the same vacuum performance and feature an IEC connector for main supply and an on/off switch for easy control. This compact dry pump offers flexibility to the system designer and end user alike with a wide range of accessories available to choose from to meet your configuration requirements. This mXDS3s model comes with a single phase 115V, 50/60Hz motor.
Catalog No. NC2888792 Supplier MARSHALL SCIENTIFIC Supplier No. EDA74402906
Add to Cart
Add to Cart
Request a Quote
The Edwards Vacuum range of small dry scroll pumps is expanding with the introduction of the compact mXDS3 and mXDS3s Dry Scroll Pumps. The new mXDS3 dry scroll pump delivers 1.77 CFM (3 m^3/h^-1) pumping speed at 50Hz or 2.06 CFM (3.5 m^3/h^-1) together with an ultimate pressure of 0.1 mbar making it well suited for backing turbomolecular pumps. The mXDS3 is the standard option supplied with no inlet valve fitted, while the mXDS3s comes factory fitted with an inlet valve featuring delay opening. The mXDS3s is also supplied with an exhaust silencer. Both offer the same vacuum performance and feature an IEC connector for main supply and an on/off switch for easy control. This compact dry pump offers flexibility to the system designer and end user alike with a wide range of accessories available to choose from to meet your configuration requirements. This mXDS3s model comes with a single phase 115V, 50/60Hz motor.