Learn More
Aviva Systems Biology Corp MXI1 Antibody - middle region (ARP31402_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31402_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MXI1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MXI1. Alternate names: MAD2, MGC43220, MXD2, MXI, bHLHc11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-54208