Learn More
Aviva Systems Biology Corp MXRA7 Antibody - N-terminal region (ARP44562_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44562_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MXRA7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MXRA7. Alternate names: FLJ41492, FLJ46603, PS1TP1, TMAP1ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-53902