Learn More
Medchemexpress LLC My-1076 | 3008262-76-5 | MFCD35444266 | 99.1% | 539.57 g·mol-1 | C29H33NO9 | 10 MG
SDP
Catalog No. 5000286798
Encompass_Preferred
MY-1076 is a small-molecule inhibitor of Yes-associated protein (YAP) used for laboratory research. It induces YAP degradation and apoptosis and shows potent antiproliferative activity against several cancer cell lines with low-nanomolar IC50s. Reported properties include CAS 3008262-76-5, formula C29H33NO9, molecular weight 539.57 g·mol-1, and high reported purity.
  • Induces YAP degradation and apoptosis in vitro
  • Potent antiproliferative activity with low-nanomolar IC50s
  • High purity suitable for analytical and biological studies
  • Available in multiple small-scale quantities for research use
  • Intended for research use only; not for human or clinical use
Catalog No. 50-002-86798 Supplier Medchemexpress LLC Supplier No. HY15536010MG
MY-1076 is a small-molecule inhibitor of Yes-associated protein (YAP) used for laboratory research. It induces YAP degradation and apoptosis and shows potent antiproliferative activity against several cancer cell lines with low-nanomolar IC50s. Reported properties include CAS 3008262-76-5, formula C29H33NO9, molecular weight 539.57 g·mol-1, and high reported purity.
  • Induces YAP degradation and apoptosis in vitro
  • Potent antiproliferative activity with low-nanomolar IC50s
  • High purity suitable for analytical and biological studies
  • Available in multiple small-scale quantities for research use
  • Intended for research use only; not for human or clinical use