Learn More
Medchemexpress LLC HY-100933 5mg Medchemexpress, MY-5445 CAS:78351-75-4 Purity:>98%
SDP
Catalog No. 501931995
Encompass_Preferred
Medchemexpress, HY-100933 5mg MY-5445 CAS:78351-75-4 MY-5445 is a specific phosphodiesterase type 5 ( PDE5 ) inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1995 Supplier Medchemexpress LLC Supplier No. HY1009335MG
Medchemexpress, HY-100933 5mg MY-5445 CAS:78351-75-4 MY-5445 is a specific phosphodiesterase type 5 ( PDE5 ) inhibitor [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.