Learn More
Medchemexpress LLC My-875 | C21H25NO6 | 5 MG
SDP
Catalog No. 5000375502
Encompass_Preferred
MY-875 is a competitive microtubulin polymerization inhibitor with an IC50 value of 0.92 μM. It inhibits microtubulin polymerization by targeting colchicine binding sites and activates the Hippo pathway, inducing apoptosis and demonstrating anticancer activity.
  • Inhibits microtubulin polymerization
  • Activates the Hippo pathway
  • Induces apoptosis
  • Exhibits anticancer activity
  • Significant anti-proliferative activity against various cancer cells
  • Arrests cells in the G2/M phase
Catalog No. 50-003-75502 Supplier Medchemexpress LLC Supplier No. HY1507615MG
MY-875 is a competitive microtubulin polymerization inhibitor with an IC50 value of 0.92 μM. It inhibits microtubulin polymerization by targeting colchicine binding sites and activates the Hippo pathway, inducing apoptosis and demonstrating anticancer activity.
  • Inhibits microtubulin polymerization
  • Activates the Hippo pathway
  • Induces apoptosis
  • Exhibits anticancer activity
  • Significant anti-proliferative activity against various cancer cells
  • Arrests cells in the G2/M phase