Learn More
Medchemexpress LLC MY-875 | C21H25NO6 | 50 MG
SDP
Catalog No. 5000375462
Encompass_Preferred
MY-875 is a competitive microtubulin polymerization inhibitor that targets colchicine binding sites and activates the Hippo pathway. This compound induces apoptosis and exhibits significant anticancer activity against various cancer cell lines.
  • Inhibits microtubulin polymerization
  • Activates the Hippo pathway
  • Induces apoptosis
  • Demonstrates anticancer activity against multiple cancer cells
  • For research use only
Catalog No. 50-003-75462 Supplier Medchemexpress LLC Supplier No. HY15076150MG
MY-875 is a competitive microtubulin polymerization inhibitor that targets colchicine binding sites and activates the Hippo pathway. This compound induces apoptosis and exhibits significant anticancer activity against various cancer cell lines.
  • Inhibits microtubulin polymerization
  • Activates the Hippo pathway
  • Induces apoptosis
  • Demonstrates anticancer activity against multiple cancer cells
  • For research use only