Learn More
Thomas Scientific MyBathT 4L Digital Water Bath, 230V
SDP
Catalog No. 501458516
Encompass_Preferred
This compact 4 liter water bath features easy to use controls that allow the user to digitally select and monitor temperature. Its hinged lid provides a secure, covered environment and can be flipped open (to the rear) or completely and instantly removed at the user's option. The MyBath is also the only water bath that comes with test tube racks for 50ml, 15ml and 1.5/2.0ml tubes, making it complete and ready to use. Designed with the molecular biology lab in mind, the MyBath 4 liter is the complete personal water bath. Digital accuracy and convenience Includes tube racks for 1.5/2.0ml, 15ml and 50ml tubes Quik-CAL™ user calibration function Hinged lid is also removable Less than 8" wide
Catalog No. 50-145-8516 Supplier Thomas Scientific Supplier No. 1213F82
This compact 4 liter water bath features easy to use controls that allow the user to digitally select and monitor temperature. Its hinged lid provides a secure, covered environment and can be flipped open (to the rear) or completely and instantly removed at the user's option. The MyBath is also the only water bath that comes with test tube racks for 50ml, 15ml and 1.5/2.0ml tubes, making it complete and ready to use. Designed with the molecular biology lab in mind, the MyBath 4 liter is the complete personal water bath. Digital accuracy and convenience Includes tube racks for 1.5/2.0ml, 15ml and 50ml tubes Quik-CAL™ user calibration function Hinged lid is also removable Less than 8" wide