Learn More
Research Products International Corp myBlock, Mini Dry Bath with Heated Lid
SDP
Catalog No. NC1778780
Encompass
The palm size Mini-Dry Bath is an exceptionally convenient block heater for use anywhere in the laboratory. Heated lid prevents evaporation and condensation. Optional 12V vehicle power adapter allows the unit to be used as a portable incubator. Various size blocks accommodate all popular size tubes from 0.2ml up to 50ml. The Mini-Dry Bath provides fast heating with accurate temperature control.
Catalog No. NC1778780 Supplier Research Products International Corp Supplier No. 400780HL
The palm size Mini-Dry Bath is an exceptionally convenient block heater for use anywhere in the laboratory. Heated lid prevents evaporation and condensation. Optional 12V vehicle power adapter allows the unit to be used as a portable incubator. Various size blocks accommodate all popular size tubes from 0.2ml up to 50ml. The Mini-Dry Bath provides fast heating with accurate temperature control.