Learn More
Mystaire™ MY Model PCR Prep Station Class 100 Enclosure Shortwave UV Bulb
SDP
Catalog No. 15338000 Shop All Mystaire Products
Change view
Click to view available options
For Use With (Equipment):
MY Model PCR Prep Station Class 100 Enclosures
Catalog No. For Use With (Equipment)
15-338-000 MY Model PCR Prep Station Class 100 Enclosures
1 options
Catalog No. 15-338-000 Supplier Mystaire™ Supplier No. MYUVB4
Add to Cart
Add to Cart
For use with MY Model PCR prep station class 100 enclosures. Effectively decontaminates the work area between amplifications.
  • 254 nanometer shortwave ultraviolet bulb
  • UV irradiation energy effectively kills bacteria and viruses on the surface of various objects
Specifications
For Use With (Equipment) MY Model PCR Prep Station Class 100 Enclosures
Length (English) 18 in.
Description 254 nm Short Wave Ultraviolet Bulb
For Use With (Application) Decontaminating Work Surfaces