Learn More
Heidolph™ Mya 4 - Reflux & Manifold Head and StarFish™ Distribution Manifold
SDP
Catalog No. 05404064 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Quick Release Male
Size:
9.5 mm
Catalog No. Type Size
05-404-064 Quick Release Male 9.5 mm
1 options
Catalog No. 05-404-064 Supplier Heidolph™ Supplier No. 015890370
Add to Cart
Add to Cart
Heidolph™ Mya 4 - Reflux & Manifold Head and StarFish™ Distribution Manifold includes shut-off
  • For use with the Mya 4 Reaction Station Kit and StarFish™ Work Station
  • Allows for the connection of either coolants, or gas/vacuum
Specifications
Description Barbed
For Use With (Equipment) Mya 4 Reaction Station Kit and StarFish™ Work Station
For Use With (Application) Allows for the Connection of Coolant or Gas/Vacuum
Includes Shut-off (6.4 mm ID)
Size 9.5 mm
Type Quick Release Male