Learn More
Aviva Systems Biology Corp MYB Antibody - N-terminal region (ARP38611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYB. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human MYB. Alternate names: Cmyb, c-myb, c-myb_CDS, efgApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56728