Learn More
Research Products International Corp MyBath 12 Liter Water Bath
SDP
Catalog No. 501976918
Encompass
The MyBath series of water baths are designed to guarantee superior accuracy and performance in a compact footprint. Easy to use controls allow the user to digitally select and monitor settings. Adjustable in 0.1°C increments, the temperature is shown on a large 3 digit display. The Quick-CAL feature allows for simple, in lab calibration. High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. Featuring a seamless, stainless steel chamber for easy cleaning and a long life. The chambers resist rust and have no corners to collect dirt or contamination.
The MyBath 12 Liter also features a drain for hassle free water changes. Units also include a see through cover that can be removed, or flipped back on a rear hinge to conserve bench space. The lid's gabled design will divert condensation to the front and back of the bath, away from the samples.
Catalog No. 50-197-6918 Supplier Research Products International Corp Supplier No. 400764
The MyBath series of water baths are designed to guarantee superior accuracy and performance in a compact footprint. Easy to use controls allow the user to digitally select and monitor settings. Adjustable in 0.1°C increments, the temperature is shown on a large 3 digit display. The Quick-CAL feature allows for simple, in lab calibration. High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. Featuring a seamless, stainless steel chamber for easy cleaning and a long life. The chambers resist rust and have no corners to collect dirt or contamination.
The MyBath 12 Liter also features a drain for hassle free water changes. Units also include a see through cover that can be removed, or flipped back on a rear hinge to conserve bench space. The lid's gabled design will divert condensation to the front and back of the bath, away from the samples.