Learn More
Thomas Scientific MyBathT 2L Digital Water Bath, 230V
SDP
Catalog No. 501458339
Encompass_Preferred
*This item has a European plug not suitable for use in the USA. Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes The myBath series is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration.* High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. All myBath models feature seamless, stainless steel chambers for easy cleaning and long life
Catalog No. 50-145-8339 Supplier Thomas Scientific Supplier No. 1200F97
*This item has a European plug not suitable for use in the USA. Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes The myBath series is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration.* High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. All myBath models feature seamless, stainless steel chambers for easy cleaning and long life