Learn More
Research Products International Corp MyBath 4L Water Bath, Complete Personal Water Bath
SDP
Catalog No. 501976970
Encompass
This compact 4 liter water bath features easy to use controls that allow the user to digitally select and monitor temperature. Its hinged lid provides a secure, covered environment and can be flipped open (towards the back) or completely and instantly removable. Designed with the molecular biology lab in mind, the MyBath 4L is the complete personal water bath.
The MyBath is also supplied with test tube racks for 41 x 1.5-2.0ml tubes, 14 x 15ml tubes or 15 x 50ml tubes making it complete and ready to use.
Catalog No. 50-197-6970 Supplier Research Products International Corp Supplier No. 400760
This compact 4 liter water bath features easy to use controls that allow the user to digitally select and monitor temperature. Its hinged lid provides a secure, covered environment and can be flipped open (towards the back) or completely and instantly removable. Designed with the molecular biology lab in mind, the MyBath 4L is the complete personal water bath.
The MyBath is also supplied with test tube racks for 41 x 1.5-2.0ml tubes, 14 x 15ml tubes or 15 x 50ml tubes making it complete and ready to use.