Learn More
Alkali Scientific MyBath B2000-12 12L Digital Wa
SDP
Catalog No. 503029611
Encompass_Preferred
The Alkali ScientificMyBath B2000-12 12L Digital Water Bath is designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a myBath to fit every lab Easy to use controls allow the user to digitaly select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch Hinged lid (included) also removable Quik-CALuser calibration function Seamless stainless steel chamber myBath 4L Includes racks for 1 5/2 0 15 50ml tubes All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The larger 8L and 12L units have drains for hassle-free water changes The 4L 8L
Catalog No. 50-302-9611 Supplier Alkali Scientific Supplier No. RS200012
The Alkali ScientificMyBath B2000-12 12L Digital Water Bath is designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a myBath to fit every lab Easy to use controls allow the user to digitaly select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch Hinged lid (included) also removable Quik-CALuser calibration function Seamless stainless steel chamber myBath 4L Includes racks for 1 5/2 0 15 50ml tubes All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The larger 8L and 12L units have drains for hassle-free water changes The 4L 8L