Learn More
Alkali Scientific Alkali Scientific™ MyBath B2000-2 2L Digital Water Bath
SDP
Catalog No. 503029665
Encompass_Preferred
The myBath series Digital Water Baths is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration. High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes
Catalog No. 50-302-9665 Supplier Alkali Scientific Supplier No. RS20002
The myBath series Digital Water Baths is designed to guarantee superior accuracy and performance in a compact footprint. With four sizes to choose from, there is a myBath to fit every lab. Easy to use controls allow the user to digitaly select and monitor settings. Adjustable in 0.1ºC increments, the temperature is shown on a large, 3-digit display. The proprietary Quik-CAL™ feature allows for simple, in-lab calibration. High efficiency insulation maintains temperature stability and uniformity within the bath, while keeping the housing cool to the touch. Hinged lid (included) also removable Quik-CAL™ user calibration function Seamless, stainless steel chamber myBath 4L Includes racks for 1.5/2.0, 15 & 50ml tubes