Learn More
Alkali Scientific MyBath B2000-4 4L Digital Wate
SDP
Catalog No. 503029616
Encompass_Preferred
Alkali ScientificMyBath (B2000-4) digital water bath are designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a MyBath to fit every lab Easy to use controls allow the user to digitally select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The 4L models includes a see-through cover that can be removed or flipped back on a rear hinge to conserve bench space The lids gabled design will divert condensation to the front and back of the bath away from samples This model offers excep
Catalog No. 50-302-9616 Supplier Alkali Scientific Supplier No. RS20004
Alkali ScientificMyBath (B2000-4) digital water bath are designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a MyBath to fit every lab Easy to use controls allow the user to digitally select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The 4L models includes a see-through cover that can be removed or flipped back on a rear hinge to conserve bench space The lids gabled design will divert condensation to the front and back of the bath away from samples This model offers excep