Learn More
Alkali Scientific MyBath B2000-8 8L Digital Wate
SDP
Catalog No. 503029667
Encompass_Preferred
The Alkali ScientificMyBath (B2000-8) digital water bath is designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a MyBath to fit every lab Easy to use controls allow the user to digitally select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The larger 8L and 12L units have drains for hassle-free water changes The 4L 8L and 12L models include a see-through cover that can be removed or flipped back on a rear hinge to conserve bench space The lids gabled design will divert conden
Catalog No. 50-302-9667 Supplier Alkali Scientific Supplier No. RS20008
The Alkali ScientificMyBath (B2000-8) digital water bath is designed to guarantee superior accuracy and performance in a compact footprint With four sizes to choose from there is a MyBath to fit every lab Easy to use controls allow the user to digitally select and monitor settings Adjustable in 0 1 C increments the temperature is shown on a large 3-digit display The proprietary Quik-CALfeature allows for simple in-lab calibration High efficiency insulation maintains temperature stability and uniformity within the bath while keeping the housing cool to the touch All Alkali ScientificMyBath models feature seamless stainless steel chambers for easy cleaning and long life The chambers resist rust and have no corners to collect dirt or contamination The larger 8L and 12L units have drains for hassle-free water changes The 4L 8L and 12L models include a see-through cover that can be removed or flipped back on a rear hinge to conserve bench space The lids gabled design will divert conden