Learn More
Alkali Scientific MyBath B2023-10 CR Circulating
SDP
Catalog No. 503029670
Encompass_Preferred
Alkalis myBathCR circulating water baths offer the same accuracy and convenience of our static myBathseries but with faster heat ramping and enhanced stability/uniformity With constant circulation water temperature is uniform throughout the chamber eliminating cold-spots and allowing for quick recovery following insertion of frozen samples The myBathCR utilizes digital PID processing control that accurately maintains and adjusts the temperature to the set parameters Once the desired temperature has been programmed the instrument begins to heat while indicating both the set and actual temperatures on the large digital display The chamber and heating coil are constructed of stainless steel resulting in easy cleaning Optional accessories are available for securing test-tubes and an optional gabled lid can be purchased to improve efficiency Constant water circulationensures even temperature distribution Digital PID processing for precise temperature control Stainless steel chamber a
Catalog No. 50-302-9670 Supplier Alkali Scientific Supplier No. CRWB2310
Alkalis myBathCR circulating water baths offer the same accuracy and convenience of our static myBathseries but with faster heat ramping and enhanced stability/uniformity With constant circulation water temperature is uniform throughout the chamber eliminating cold-spots and allowing for quick recovery following insertion of frozen samples The myBathCR utilizes digital PID processing control that accurately maintains and adjusts the temperature to the set parameters Once the desired temperature has been programmed the instrument begins to heat while indicating both the set and actual temperatures on the large digital display The chamber and heating coil are constructed of stainless steel resulting in easy cleaning Optional accessories are available for securing test-tubes and an optional gabled lid can be purchased to improve efficiency Constant water circulationensures even temperature distribution Digital PID processing for precise temperature control Stainless steel chamber a