Learn More
Sigma Aldrich Fine Chemicals Biosciences MyBathTM Digital Water Bat1EA
Catalog No. 502908926
Encompass
MyBath water bath features easy to use controls that allow the user to digitally select and monitor temperatureThe temperature is shown on a large 3-digit display Accuracy /- 0.5C. Increments 0.1C. Uniformity /- 0.2CTest tube rack inserts are sold separatelyHinged lid provides a secure covered environment and can be flipped open (to the rear) or completely and instantly removedDrain allows for hassle-free water changesQuik-CAL user calibration functionSeamless stainless steel chamber the chambers resist rust and have no corners to collect dirt or contamination
Catalog No. 50-290-8926 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSB200081EA
MyBath water bath features easy to use controls that allow the user to digitally select and monitor temperatureThe temperature is shown on a large 3-digit display Accuracy /- 0.5C. Increments 0.1C. Uniformity /- 0.2CTest tube rack inserts are sold separatelyHinged lid provides a secure covered environment and can be flipped open (to the rear) or completely and instantly removedDrain allows for hassle-free water changesQuik-CAL user calibration functionSeamless stainless steel chamber the chambers resist rust and have no corners to collect dirt or contamination