Learn More
Sigma Aldrich Fine Chemicals Biosciences MyBathTM mini water bath A1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7414211EA
Encompass
Designed with the molecular biology lab in mind the MyBath(TM) mini water bath system is the complete personal water bath. This compact water bath features easy to use controls that allow the user to digitally select and monitor temperature.Its hinged lid provides a secure covered environment and can be flipped open (to the rear) or completely and instantly removed at the users option. It includes Quik-Cal(TM) in-lab user calibration function.Note the racks only comes with the 4L model
Catalog No. 50-290-9099