Learn More
Aviva Systems Biology Corp MYBBP1A Antibody - C-terminal region (ARP37224_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37224_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MYBBP1A. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of mouse MYBBP1A. Alternate names: P160, p67MBP, p160MBP, AL024407, AU019902ApplicationIHC, WB. Species reactivity: Guinea Pig, Mouse, Rat
Catalog No. 50-014-56339