Learn More
Aviva Systems Biology Corp MYBBP1A Antibody - C-terminal region (ARP39099_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39099_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MYBBP1A. It was validated on Western Blot. . Alternate names: FLJ37886, P160, PAP2ApplicationWB. Species reactivity: Human
Catalog No. 50-284-781