Learn More
Aviva Systems Biology Corp Mybbp1a Antibody - C-terminal region (ARP37905_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37905_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Mybbp1a. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse Mybbp1a. Alternate names: AL024407, AU019902, P160, p160MBP, p67MBPApplicationWB. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-56460