Learn More
Aviva Systems Biology Corp MYBL1 Antibody - middle region (ARP32707_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32707_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MYBL1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human MYBL1. Alternate names: A-MYB, AMYB, MGC120059, MGC120061ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54716