Learn More
Research Products International Corp myBlock, Mini Dry Bath with Cooling
SDP
Supplier:  Research Products International Corp 400780C
Encompass
The palm size Mini-Dry Bath is an exceptionally convenient block heater for use anywhere in the laboratory. Optional 12V vehicle power adapter allows the unit to be used as a portable incubator. Various size blocks accommodate all popular size tubes from 0.2ml up to 50ml. The Mini-Dry Bath provides fast heating with accurate temperature control.
Catalog No. 50-222-9548