Learn More
Thomas Scientific MyBlockT Mini Dry Bath with Heated Lid, 100-240V (US Plug)
SDP
Catalog No. 501460133
Encompass_Preferred
The new MyBlock™ HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation. Both the block and lid temperature can be selected independently in increments of 0.1°C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures. Heated lid prevents evaporation & condensation Compact, fits in the palm of your hand Independant temperature control of lid & block Exchangeable blocks for 0.2 to 0.5ml tubes Room temperature to 37°C in just 2 minutes
Catalog No. 50-146-0133 Supplier Thomas Scientific Supplier No. 1213F91
The new MyBlock™ HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation. Both the block and lid temperature can be selected independently in increments of 0.1°C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions & other temperature hyper-sensitive procedures. Heated lid prevents evaporation & condensation Compact, fits in the palm of your hand Independant temperature control of lid & block Exchangeable blocks for 0.2 to 0.5ml tubes Room temperature to 37°C in just 2 minutes