Learn More
Sigma Aldrich Fine Chemicals Biosciences myBlock ll digital dry bat1EA
Catalog No. 502908670
Encompass
myBlock series is a next generation product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls timed or continuous operation and a removable hinged lid. In addition an optional external temperature feedback probe can be positioned directly inside a sample tube providing real-time control and monitoring of actual sample temperature. MyBlocks transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. The hinged lid can be flipped open (to the rear) or easily removed (lifted off) to accommodate taller tubes.Digital temperature control (set and walk away)Hinged lid (improves uniformity conserves energy)
Catalog No. 50-290-8670 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BMSBSH50022B1EA
myBlock series is a next generation product line that introduces several major advantages over traditional dry baths. They are the first digital dry baths to include advanced microprocessor controls timed or continuous operation and a removable hinged lid. In addition an optional external temperature feedback probe can be positioned directly inside a sample tube providing real-time control and monitoring of actual sample temperature. MyBlocks transparent lid helps to promote better uniformity and minimize temperature stratification in the block by maintaining a warm air jacket layer. The hinged lid can be flipped open (to the rear) or easily removed (lifted off) to accommodate taller tubes.Digital temperature control (set and walk away)Hinged lid (improves uniformity conserves energy)