Learn More
Braintree Scientific MyBlock Mini Digital Dry Bath, blocks sold separately
SDP
Catalog No. 501954397
Encompass
Benchtop control of sample temperature has never been this easy or economical. MyBlock Mini fits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 4.4 inches wide, MyBlock Mini is truly the first personal block incubator. Compact, fits in the palm of your hand. Exchangeable blocks, for tubes .2 to 50ml. Clear hinged cover ensures temperature uniformity. Technical Data: Temperature range: Ambient +5 to 100C; Temperature accuracy: 0.5C; Temperature increments: .1C; Temperature uniformity: 0.2C; Dimensions (WxDxH): 4.4" x 5.9" x 4", 11cm x 15cm x 10cm. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4397 Supplier Braintree Scientific Supplier No. BSH200120V
Benchtop control of sample temperature has never been this easy or economical. MyBlock Mini fits almost anywhere and can even be used "on-the-go" in cars, boats or wherever a 12 volt power source is available. Its simple touch pad control with digital display is designed for "set and walk away" temperature selection and unrivaled accuracy. At less than 4.4 inches wide, MyBlock Mini is truly the first personal block incubator. Compact, fits in the palm of your hand. Exchangeable blocks, for tubes .2 to 50ml. Clear hinged cover ensures temperature uniformity. Technical Data: Temperature range: Ambient +5 to 100C; Temperature accuracy: 0.5C; Temperature increments: .1C; Temperature uniformity: 0.2C; Dimensions (WxDxH): 4.4" x 5.9" x 4", 11cm x 15cm x 10cm. **Sold for research purposes only and not for human use**