Learn More
Braintree Scientific MyBlock Mini Digital Dry Bath w/heated lid, blocks sold sep.
SDP
Catalog No. 501954403
Encompass
The MyBlock™ HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation. Both the block and lid temperature can be selected independently in increments of .1C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions and other temperature hyper-sensitive procedures. Heated lid prevents evaporation and condensation. Compact, fits in the palm of user's hand. Independant temperature control of lid and block. Exchangeable blocks for 0.2 to 0.5ml tubes. Room temperature to 37C in just 2 minutes. Specifications: Temperature range: Ambient +5 to 100C; Temperature accuracy: .3C; Temperature increments: .1C; Temperature uniformity: .2C; Dimensions: 4" x 6" x 4.5"/10.5cm x 15cm x 11.5cm. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4403 Supplier Braintree Scientific Supplier No. BSH200HL115V
The MyBlock™ HL is the first and only mini heat block featuring a heated lid for protecting samples against unwanted condensation and evaporation. Both the block and lid temperature can be selected independently in increments of .1C, making this MyBlock the ideal choice for the most temperature sensitive applications, including immunoassays, LAMP assays, enzyme reactions and other temperature hyper-sensitive procedures. Heated lid prevents evaporation and condensation. Compact, fits in the palm of user's hand. Independant temperature control of lid and block. Exchangeable blocks for 0.2 to 0.5ml tubes. Room temperature to 37C in just 2 minutes. Specifications: Temperature range: Ambient +5 to 100C; Temperature accuracy: .3C; Temperature increments: .1C; Temperature uniformity: .2C; Dimensions: 4" x 6" x 4.5"/10.5cm x 15cm x 11.5cm. **Sold for research purposes only and not for human use**